Ethyl-2,3-dihydroxybenzoat
Lieferant: Apollo Scientific
OR25996-1G
APOSOR25996-1GEA
105
EUR
APOSOR25996-1G
Ethyl-2,3-dihydroxybenzoat
Ethyl-2,3-dihydroxybenzoat
Formel:
C₉H₁₀O₄ Molecular Weight: 182,18 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03644149 CAS-Nummer: 3943-73-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...