2-Brom-4-methylthiazol

Lieferant: Apollo Scientific
Achtung

OR2600-1G OR2600-5G
APOSOR2600-1GEA 21.9 EUR
APOSOR2600-1G APOSOR2600-5G
2-Brom-4-methylthiazol
2-Bromo-4-methylthiazole
Formel: C₄H₄BrNS
Molecular Weight: 178.05 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06660130
CAS-Nummer: 7238-61-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR