2-Brom-4-methylthiazol
Lieferant: Apollo Scientific
|
Achtung
|
OR2600-1G
OR2600-5G
APOSOR2600-1GEA
21.9
EUR
APOSOR2600-1G
APOSOR2600-5G
2-Brom-4-methylthiazol
2-Bromo-4-methylthiazole
Formel:
C₄H₄BrNS Molecular Weight: 178.05 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD06660130 CAS-Nummer: 7238-61-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...