6-Aminobenzo[b]thiophene 1,1-dioxide

Lieferant: Apollo Scientific
Achtung

Synonyme: 6-Amino-1,1-dioxobenzo[b]thiophene, 6-Amino-1,1-dioxo-1-benzothiophene

OR26007-1G
APOSOR26007-1GEA 121 EUR
APOSOR26007-1G
6-Aminobenzo[b]thiophene 1,1-dioxide
6-Aminobenzo[b]thiophene 1,1-dioxide
Formel: C₈H₇NO₂S
Molecular Weight: 181.212 g/mol
Schmelzpunkt: 150…156 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173800
CAS-Nummer: 20503-40-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR