5-Chlorobenzo[b]thiophene-3-acetic acid
Lieferant: Apollo Scientific
Synonyme:
2-(5-Chlorobenzo[b]thiophen-3-yl)acetic acid
OR26012-1G
OR26012-250MG
APOSOR26012-1GEA
0
EUR
APOSOR26012-1G
APOSOR26012-250MG
5-Chlorobenzo[b]thiophene-3-acetic acid
5-Chlorobenzo[b]thiophene-3-acetic acid
Formel:
C₁₀H₇ClO₂S Molecular Weight: 226.68 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052308 CAS-Nummer: 17266-30-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...