5-Chlorobenzo[b]thiophene-3-acetic acid

Lieferant: Apollo Scientific

OR26012-1G OR26012-250MG
APOSOR26012-1GEA 0 EUR
APOSOR26012-1G APOSOR26012-250MG
5-Chlorobenzo[b]thiophene-3-acetic acid
5-Chlorobenzo[b]thiophene-3-acetic acid
Formel: C₁₀H₇ClO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00052308
CAS-Nummer: 17266-30-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR