2-Bromo-1-(3-methylbenzo[b]thiophen-2-yl)ethan-1-one

Lieferant: Apollo Scientific
Gefahr

Synonyme: 2-(Bromoacetyl)-3-methylbenzo[b]thiophene

OR26021-250MG
APOSOR26021-250MGEA 87 EUR
APOSOR26021-250MG
2-Bromo-1-(3-methylbenzo[b]thiophen-2-yl)ethan-1-one
2-Bromo-1-(3-methylbenzo[b]thiophen-2-yl)ethan-1-one
Formel: C₁₁H₉BrOS
Molecular Weight: 269,17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00173793
CAS-Nummer: 118337-33-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR