(5-Methyl-1,3-thiazol-2-yl)methanol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(Hydroxymethyl)-5-methyl-1,3-thiazole

OR2604-250MG
APOSOR2604-250MGEA 157 EUR
APOSOR2604-250MG
(5-Methyl-1,3-thiazol-2-yl)methanol
(5-Methyl-1,3-thiazol-2-yl)methanol
Formel: C₅H₇NOS
Molecular Weight: 129,18 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD08060774
CAS-Nummer: 202932-04-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR