2-[4-(Methanesulphonyloxy)phenyl]acetic acid

Lieferant: Apollo Scientific
Gefahr

OR2610-250MG
APOSOR2610-250MGEA 114 EUR
APOSOR2610-250MG
2-[4-(Methanesulphonyloxy)phenyl]acetic acid
2-[4-(Methanesulphonyloxy)phenyl]acetic acid
Formel: C₉H₁₀O₅S
Molecular Weight: 230,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09800580
CAS-Nummer: 64369-79-5
UN: 1759
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR