1-(4-Methoxybenzyl)-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

OR2611-500MG
APOSOR2611-500MGEA 0 EUR
APOSOR2611-500MG
1-(4-Methoxybenzyl)-5-oxopyrrolidine-3-carboxylic acid
1-(4-Methoxybenzyl)-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₃H₁₅NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD03834522
CAS-Nummer: 96449-89-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR