2,6-Dimethoxy-3-pyridinboronsäure

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid

OR2616-250MG
APOSOR2616-250MGEA 21.9 EUR
APOSOR2616-250MG
2,6-Dimethoxy-3-pyridinboronsäure
2,6-Dimethoxy-3-pyridineboronic acid
Formel: C₇H₁₀BNO₄
Molecular Weight: 182,97 g/mol
Schmelzpunkt: 100…117 °C
Lagertemperatur: Kühlschrank
MDL: MFCD03788239
CAS-Nummer: 221006-70-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR