5-Methoxy-2-nitrophenol 97%
Lieferant: Apollo Scientific
|
Achtung
|
OR26237-25G
APOSOR26237-25GEA
128
EUR
APOSOR26237-25G
5-Methoxy-2-nitrophenol 97%
5-Methoxy-2-nitrophenol
Formel:
C₇H₇NO₄ Schmelzpunkt: 92…94 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00100932 CAS-Nummer: 704-14-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...