Indan-1-one oxime

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-1H-inden-1-one oxime, 1-(Hydroxyimino)indane

OR26239-1G
APOSOR26239-1GEA 26.1 EUR
APOSOR26239-1G
Indan-1-one oxime
Indan-1-one oxime
Formel: C₉H₉NO
Molecular Weight: 147.174 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00082974
CAS-Nummer: 3349-60-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR