Bicyclo[2.2.1]heptan-2-ol 95%

Lieferant: Apollo Scientific

OR26341-5G
APOSOR26341-5GEA 120 EUR
APOSOR26341-5G
Bicyclo[2.2.1]heptan-2-ol 95%
Bicyclo[2.2.1]heptan-2-ol
Formel: C₇H₁₂O
Molecular Weight: 112.17 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00167747
CAS-Nummer: 1632-68-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR