2,4-Dichloro-8-methoxy-3-phenylquinoline

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dichloro-8-methoxy-3-phenyl-1-azanaphthalene

OR26411-100MG
APOSOR26411-100MGEA 21.9 EUR
APOSOR26411-100MG
2,4-Dichloro-8-methoxy-3-phenylquinoline
2,4-Dichloro-8-methoxy-3-phenylquinoline
Formel: C₁₆H₁₁Cl₂NO
Molecular Weight: 304,171 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00101113
CAS-Nummer: 274691-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR