6-Amino-2-thioxo-2,3-dihydro-4(1H)-pyrimidinonhydrat

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-6-hydroxy-2-mercaptopyrimidine monohydrate, 4-Amino-6-hydroxy-2-mercaptopyrimidine Hydrate, 6-Amino-2-thiouracil hydrate, 6-Amino-2-thiouracil Hydrate

OR2651-5G
APOSOR2651-5GEA 21.9 EUR
APOSOR2651-5G
6-Amino-2-thioxo-2,3-dihydro-4(1H)-pyrimidinonhydrat
4-Amino-6-hydroxy-2-mercaptopyrimidine hydrate
Formel: C₄H₅N₃OS·xH₂O
Molecular Weight: 143,17 g/mol (anhy)
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052384
CAS-Nummer: 65802-56-4
EINECS: 213-722-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR