Ethyl phenylsulphonylacetate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 2-(phenylsulphonyl)acetate, Phenylsulphonylacetic acid Ethyl Ester

OR26538-1G
APOSOR26538-1GEA 21.9 EUR
APOSOR26538-1G
Ethyl phenylsulphonylacetate
Ethyl phenylsulphonylacetate
Formel: C₁₀H₁₂O₄S
Molecular Weight: 228,26 g/mol
Siedepunkt: 135 °C
Schmelzpunkt: 44 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00025041
CAS-Nummer: 7605-30-3
EINECS: 231-516-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR