Thiophene-3,4-dicarbonitrile

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,4-Dicyanothiophene

OR2655-5G
APOSOR2655-5GEA 97.3 EUR
APOSOR2655-5G
Thiophene-3,4-dicarbonitrile
Thiophene-3,4-dicarbonitrile
Formel: C₆H₂N₂S
Molecular Weight: 134,158 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01233047
CAS-Nummer: 18853-32-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR