2-[(4-Nitrophenoxy)methyl]oxirane

Lieferant: Apollo Scientific
Achtung

OR26560-1G OR26560-250MG
APOSOR26560-1GEA 69.6 EUR
APOSOR26560-1G APOSOR26560-250MG
2-[(4-Nitrophenoxy)methyl]oxirane
2-[(4-Nitrophenoxy)methyl]oxirane
Formel: C₉H₉NO₄
Molecular Weight: 195.18 g/mol
Schmelzpunkt: 62…68 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00041845
CAS-Nummer: 5255-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR