2-Chloro-4-iodopyridine-3-carboxaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-4-iodonicotinaldehyde
OR2658-1G
APOSOR2658-1GEA
21.9
EUR
APOSOR2658-1G
2-Chloro-4-iodopyridine-3-carboxaldehyde
2-Chloro-4-iodopyridine-3-carboxaldehyde
Formel:
C₆H₃ClINO Molecular Weight: 267,45 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD03095294 CAS-Nummer: 153034-90-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...