Methyl 2-bromomethyl-4-cyanobenzoate 95+%

Lieferant: Apollo Scientific

Synonyme: Methyl 2-(bromomethyl)-4-cyanobenzoate

OR2663-1G
APOSOR2663-1GEA 252 EUR
APOSOR2663-1G
Methyl 2-bromomethyl-4-cyanobenzoate 95+%
Methyl 2-bromomethyl-4-cyanobenzoate
Formel: C₁₀H₈BrNO₂
Molecular Weight: 254,08 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD09864855
CAS-Nummer: 165111-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR