1-(Aminomethyl)cyclohexane-1-carboxylic acid, N-BOC protected

Lieferant: Apollo Scientific
Achtung

OR2665-1G
APOSOR2665-1GEA 0 EUR
APOSOR2665-1G
1-(Aminomethyl)cyclohexane-1-carboxylic acid, N-BOC protected
1-(Aminomethyl)cyclohexane-1-carboxylic acid, N-BOC protected
Formel: C₁₃H₂₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD09264198
CAS-Nummer: 204514-23-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR