[4-({{(9H-Fluoren-9-yl)methoxy]carbonyl}amino)tetrahydro-2H-pyran-4-yl]acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Amino-4-(carboxymethyl)tetrahydro-2H-pyran, N-FMOC protected, (9H-Fluoren-9-yl)methyl [4-(carboxymethyl)tetrahydro-2H-pyran-4-yl]carbamate

OR2670-1G
APOSOR2670-1GEA 557 EUR
APOSOR2670-1G
[4-({{(9H-Fluoren-9-yl)methoxy]carbonyl}amino)tetrahydro-2H-pyran-4-yl]acetic acid
[4-({{(9H-Fluoren-9-yl)methoxy]carbonyl}amino)tetrahydro-2H-pyran-4-yl]acetic acid
Formel: C₂₂H₂₃NO₅
Molecular Weight: 381,422 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09264206
CAS-Nummer: 946716-25-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR