[4-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)tetrahydro-2H-thiopyran-4-yl]acetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: (9H-Fluoren-9-yl)methyl [4-(carboxymethyl)tetrahydro-2H-thiopyran-4-yl]carbamate, 4-Amino-4-(carboxymethyl)tetrahydro-2H-thiopyran, N-FMOC protected

OR2672-100MG
APOSOR2672-100MGEA 48 EUR
APOSOR2672-100MG
[4-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)tetrahydro-2H-thiopyran-4-yl]acetic acid
[4-({[(9H-Fluoren-9-yl)methoxy]carbonyl}amino)tetrahydro-2H-thiopyran-4-yl]acetic acid
Formel: C₂₂H₂₃NO₄S
Molecular Weight: 397,487 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD09264207
CAS-Nummer: 946727-62-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR