Fmoc-Cyclopropylglycine

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-Amino(cyclopropyl)acetic acid, N-FMOC protected, Fmoc-L-Cyclopropylglycine

OR2679-250MG
APOSOR2679-250MGEA 77 EUR
APOSOR2679-250MG
Fmoc-Cyclopropylglycine
Fmoc-Cyclopropylglycine
Formel: C₂₀H₁₉NO₄
Molecular Weight: 337,4 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD06659145
CAS-Nummer: 1212257-18-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR