Methylchinoxalin-6-carboxylat
Lieferant: Apollo Scientific
Synonyme:
Methyl 6-Quinoxalinecarboxylate
OR26804-10G
APOSOR26804-10GEA
92.8
EUR
APOSOR26804-10G
Methylchinoxalin-6-carboxylat
Methylchinoxalin-6-carboxylat
Formel:
C₁₀H₈N₂O₂ Molecular Weight: 188,19 g/mol Schmelzpunkt: 101…102 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00102618 CAS-Nummer: 23088-23-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...