5-Methyl-2,1,3-benzothiadiazol
Lieferant: Apollo Scientific
|
Achtung
|
OR26809-250MG
APOSOR26809-250MGEA
21.9
EUR
APOSOR26809-250MG
5-Methyl-2,1,3-benzothiadiazol
5-Methyl-2,1,3-benzothiadiazole
Formel:
C₇H₆N₂S Siedepunkt: 62…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173945 CAS-Nummer: 1457-93-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...