6-Bromo-4,5-dimethyl-2,1,3-benzothiadiazole
Lieferant: Apollo Scientific
|
Achtung
|
OR26817-1G
APOSOR26817-1GEA
255
EUR
APOSOR26817-1G
6-Bromo-4,5-dimethyl-2,1,3-benzothiadiazole
6-Bromo-4,5-dimethyl-2,1,3-benzothiadiazole
Formel:
C₈H₇BrN₂S Schmelzpunkt: 84…85 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00126470 CAS-Nummer: 175204-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...