Ethyl-β-alaninathydrochlorid

Lieferant: Apollo Scientific
Achtung

Synonyme: beta-Alanine ethyl ester hydrochloride, H-beta-Ala-OEt.HCl, Ethyl 3-Aminopropionate Hydrochloride, β-Alanine ethyl ester hydrochloride, 3-Aminopropionic Acid Ethyl Ester Hydrochloride, Ethyl 3-aminopropanoate hydrochloride, 3-Aminopropionic acid Ethyl Ester Hydrochloride, H-β-Ala-OEt·HCl, Ethyl 3-aminopropionate HCl

OR26826-10G
APOSOR26826-10GEA 21.9 EUR
APOSOR26826-10G
Ethyl-β-alaninathydrochlorid
Ethyl-3-aminopropanoate hydrochloride
Formel: C₅H₁₁NO₂·HCl
Molecular Weight: 153,6 g/mol
Schmelzpunkt: 67…72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012909
CAS-Nummer: 4244-84-2
EINECS: 224-203-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR