Methyl 2-(4-chlorophenoxy)-2-methylpropanoate

Lieferant: Apollo Scientific

OR26848-1G
APOSOR26848-1GEA 87 EUR
APOSOR26848-1G
Methyl 2-(4-chlorophenoxy)-2-methylpropanoate
Methyl 2-(4-chlorophenoxy)-2-methylpropanoate
Formel: C₁₁H₁₃ClO₃
Molecular Weight: 228,672 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00203861
CAS-Nummer: 55162-41-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR