Ethyl 5-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate ≥96%

Lieferant: Apollo Scientific

OR2689-250MG
APOSOR2689-250MGEA 166 EUR
APOSOR2689-250MG
Ethyl 5-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate ≥96%
Ethyl 5-chloro-2-methylimidazo[1,2-a]pyridine-3-carboxylate
Formel: C₁₁H₁₁ClN₂O₂
Molecular Weight: 238,67 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD07366504
CAS-Nummer: 141517-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR