2-Methoxy-4-[(E)-2-nitrovinyl]phenol 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Hydroxy-5-[(E)-2-nitroethenyl]anisole, trans-4-Hydroxy-3-methoxy-β-nitrostyrene

OR26919-25G
APOSOR26919-25GEA 168 EUR
APOSOR26919-25G
2-Methoxy-4-[(E)-2-nitrovinyl]phenol 98%
2-Methoxy-4-[(E)-2-nitrovinyl]phenol
Formel: C₉H₉NO₄
Molecular Weight: 195,172 g/mol
Schmelzpunkt: 168…170 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00024827
CAS-Nummer: 6178-42-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR