4-Chloro-2-[(dimethylhydrazono)methyl]phenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(5-Chloro-2-hydroxybenzylidene)-1,1-dimethylhydrazine, 4-Chloro-2-((2,2-dimethylhydrazineylidene)methyl)phenol, 4-Chloro-2-[(dimethylhydrazinylidene)methyl]phenol
OR26981-250MG
APOSOR26981-250MGEA
52.2
EUR
APOSOR26981-250MG
4-Chloro-2-[(dimethylhydrazono)methyl]phenol
4-Chloro-2-[(dimethylhydrazono)methyl]phenol
Formel:
C₉H₁₁ClN₂O Molecular Weight: 198,649 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00662300 CAS-Nummer: 284489-44-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...