4,6-Dimethylpyridin-2-ol

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,4-Dimethyl-6-hydroxypyridine

OR27029-1G
APOSOR27029-1GEA 27.7 EUR
APOSOR27029-1G
4,6-Dimethylpyridin-2-ol
4,6-Dimethylpyridin-2-ol
Formel: C₇H₉NO
Molecular Weight: 123,16 g/mol
Schmelzpunkt: 178…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00831528
CAS-Nummer: 16115-08-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR