2,6-Dichloro-4-isocyanatopyridine
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
2,6-Dichloropyridin-4-yl isocyanate
OR27051-1G
APOSOR27051-1GEA
125
EUR
APOSOR27051-1G
2,6-Dichloro-4-isocyanatopyridine
2,6-Dichloro-4-isocyanatopyridine
When melted, may contain small amount of sediment.
Formel:
C₆H₂Cl₂N₂O Molecular Weight: 189 g/mol Siedepunkt: 105…108 °C Schmelzpunkt: 25,8…26.8 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00052828 CAS-Nummer: 159178-03-7 UN: 2206 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...