3-(2,6-Dichlorophenyl)-4-(hydroxymethyl)-5-methylisoxazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
[3-(2,6-Dichlorophenyl)-5-methylisoxazol-4-yl]methanol
OR27067-250MG
APOSOR27067-250MGEA
140
EUR
APOSOR27067-250MG
3-(2,6-Dichlorophenyl)-4-(hydroxymethyl)-5-methylisoxazole
3-(2,6-Dichlorophenyl)-4-(hydroxymethyl)-5-methylisoxazole
Formel:
C₁₁H₉Cl₂NO₂ Molecular Weight: 258,101 g/mol Schmelzpunkt: 110 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052550 CAS-Nummer: 175204-38-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...