Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate

Lieferant: Apollo Scientific
Achtung

Synonyme: Ethyl 3-cyano-2-hydroxy-6-methylisonicotinate

OR27078-1G
APOSOR27078-1GEA 46.4 EUR
APOSOR27078-1G
Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate
Ethyl-3-cyano-2-hydroxy-6-methylisonicotinate
Formel: C₁₀H₁₀N₂O₃
Molecular Weight: 206,2 g/mol
Schmelzpunkt: 218…220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052274
CAS-Nummer: 18619-97-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR