5-Chloro-2-hydroxy-4,6-dimethylnicotinonitrile

Lieferant: Apollo Scientific
Achtung

OR27087-1G
APOSOR27087-1GEA 133 EUR
APOSOR27087-1G
5-Chloro-2-hydroxy-4,6-dimethylnicotinonitrile
5-Chloro-2-hydroxy-4,6-dimethylnicotinonitrile
Formel: C₈H₇ClN₂O
Lagertemperatur: Raumtemperatur
MDL: MFCD00052652
CAS-Nummer: 23819-92-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR