2,6-Dichloro-4-methylnicotinamide
Lieferant: Apollo Scientific
|
Achtung
|
OR27097-1G
APOSOR27097-1GEA
34.8
EUR
APOSOR27097-1G
2,6-Dichloro-4-methylnicotinamide
2,6-Dichloro-4-methylnicotinamide
Formel:
C₇H₆Cl₂N₂O Siedepunkt: 129…132 °C/7mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052636 CAS-Nummer: 38841-54-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...