Ethyl 2-chloro-3-cyano-6-methylisonicotinate

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Chloro-4-(ethoxycarbonyl)-6-methylnicotinonitrile, Ethyl-2-chloro-3-cyano-6-methylisonicotinate, Ethyl 2-chloro-3-cyano-6-methylpyridine-4-carboxylate

OR27103-250MG
APOSOR27103-250MGEA 49.3 EUR
APOSOR27103-250MG
Ethyl 2-chloro-3-cyano-6-methylisonicotinate
Ethyl 2-chloro-3-cyano-6-methylisonicotinate
Formel: C₁₀H₉ClN₂O₂
Molecular Weight: 224,644 g/mol
Schmelzpunkt: 58…60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00023416
CAS-Nummer: 40108-12-1
UN: 3439
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR