1-(4,6-Dimethylpyrimidin-2-yl)pyrrolidine-2-carboxylic acid

Lieferant: Apollo Scientific

OR2715-1G
APOSOR2715-1GEA 201 EUR
APOSOR2715-1G
1-(4,6-Dimethylpyrimidin-2-yl)pyrrolidine-2-carboxylic acid
1-(4,6-Dimethylpyrimidin-2-yl)pyrrolidine-2-carboxylic acid
Formel: C₁₁H₁₅N₃O₂
Schmelzpunkt: 108…109 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD07021590
CAS-Nummer: 956614-59-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR