Pivalamidine hydrochloride

Lieferant: Apollo Scientific
Achtung

OR27177-25G
APOSOR27177-25GEA 150 EUR
APOSOR27177-25G
Pivalamidine hydrochloride
Pivalamidine hydrochloride
Formel: C₅H₁₂N₂
Schmelzpunkt: 184…186 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051988
CAS-Nummer: 18202-73-8
EINECS: 242-091-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR