4-(tert-Butyl)-2-chloranilin
Lieferant: Apollo Scientific
|
Gefahr
|
OR27179-250MG
APOSOR27179-250MGEA
42.2
EUR
APOSOR27179-250MG
4-(tert-Butyl)-2-chloranilin
4-(tert-Butyl)-2-chloranilin
Formel:
C₁₀H₁₄ClN Siedepunkt: 84 °C/0,1mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052272 CAS-Nummer: 42265-67-8 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...