Benzamidoxim

Lieferant: Apollo Scientific
Gefahr

OR27223-5G OR27223-1G OR27223-10G
APOSOR27223-5GEA 0 EUR
APOSOR27223-5G APOSOR27223-1G APOSOR27223-10G
Benzamidoxim
Benzamidoxim
Formel: C₇H₈N₂O
Schmelzpunkt: 60 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00031485
CAS-Nummer: 613-92-3
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR