2-Bromo-N-(2,4-dichlorophenyl)acetamide

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(Bromoacetyl)-2,4-dichloroaniline

OR27238-250MG
APOSOR27238-250MGEA 26.1 EUR
APOSOR27238-250MG
2-Bromo-N-(2,4-dichlorophenyl)acetamide
2-Bromo-N-(2,4-dichlorophenyl)acetamide
Formel: C₈H₆BrCl₂NO
Molecular Weight: 282,96 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD02089952
CAS-Nummer: 286841-26-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR