4-Methyl-6-(methylthio)-1,3,5-triazin-2-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-methyl-6-(methylsulphanyl)-1,3,5-triazine

OR27304-1G
APOSOR27304-1GEA 0 EUR
APOSOR27304-1G
4-Methyl-6-(methylthio)-1,3,5-triazin-2-amine
4-Methyl-6-(methylthio)-1,3,5-triazin-2-amine
Formel: C₅H₈N₄S
Molecular Weight: 156,21 g/mol
Schmelzpunkt: 152 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00052777
CAS-Nummer: 27622-90-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR