4-Isopropyl-6-(methylthio)-1,3,5-triazin-2-amine

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-isopropyl-6-(methylthio)-1,3,5-triazine

OR27306-1G
APOSOR27306-1GEA 91.5 EUR
APOSOR27306-1G
4-Isopropyl-6-(methylthio)-1,3,5-triazin-2-amine
4-Isopropyl-6-(methylthio)-1,3,5-triazin-2-amine
Formel: C₇H₁₂N₄S
Molecular Weight: 184.26 g/mol
Schmelzpunkt: 127…130 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052790
CAS-Nummer: 175204-55-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR