4-(Methylthio)-6-(2-thienyl)-1,3,5-triazin-2-amine
Lieferant: Apollo Scientific
|
Achtung
|
OR27312-1G
APOSOR27312-1GEA
95.7
EUR
APOSOR27312-1G
4-(Methylthio)-6-(2-thienyl)-1,3,5-triazin-2-amine
4-(Methylthio)-6-(2-thienyl)-1,3,5-triazin-2-amine
Formel:
C₈H₈N₄S₂ Molecular Weight: 224,306 g/mol Schmelzpunkt: 176…177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052772 CAS-Nummer: 175204-61-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...