4-Amino-6-methyl-1,3,5-triazin-2-ol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-Amino-4-hydroxy-6-methyl-1,3,5-triazine

OR27314-250MG
APOSOR27314-250MGEA 21.9 EUR
APOSOR27314-250MG
4-Amino-6-methyl-1,3,5-triazin-2-ol 95%
4-Amino-6-methyl-1,3,5-triazin-2-ol
Formel: C₄H₆N₄O
Molecular Weight: 126,1200 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052767
CAS-Nummer: 16352-06-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR