4-(1-Adamantyl)-6-amino-1,3,5-trazin-2-ol
Lieferant: Apollo Scientific
Synonyme:
2-(Adamant-1-yl)-4-amino-6-hydroxy-1,3,5-triazine
OR27324-1G
APOSOR27324-1GEA
248
EUR
APOSOR27324-1G
4-(1-Adamantyl)-6-amino-1,3,5-trazin-2-ol
4-(1-Adamantyl)-6-amino-1,3,5-trazin-2-ol
Formel:
C₁₃H₁₈N₄O Molecular Weight: 246,31 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078504 CAS-Nummer: 151250-94-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...